missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, Dihydrate, Crystal, Reagent, 99-102%, Spectrum™ Chemical

Catalog No. 18605565
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605565 25 g Poly Bottle
18605566 125 g Poly Bottle
18605567 500 g Poly Bottle
18605569 2.5 kg Poly Bottle
18605568 6 x 500 g Poly Bottle
18605570 12 kg Poly Pail
18605571 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18605565 Supplier Spectrum Chemical Mfg Cor Supplier No. P131025GM
Get Up to $500 in Chemicals with Purchase
View all promotions

P1310, 13455-21-5, KF·2H2O

Spectrum™ Chemical Potassium Fluoride, Dihydrate, Crystal, Reagent is a source of fluoride, it is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
IUPAC Name potassium dihydrate fluoride
SMILES O.O.[F-].[K+]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99 to 102%
Grade Reagent
Packaging Poly Bottle