missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride, Dihydrate, Crystal, Reagent, 99-102%, Spectrum™ Chemical
P1310, 13455-21-5, KF·2H2O
Supplier: Spectrum Chemical Mfg Cor P1310500GM
Description
Spectrum™ Chemical Potassium Fluoride, Dihydrate, Crystal, Reagent is a source of fluoride, it is the preferred reagent in the conversion process of chlorocarbons into fluorocarbons. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.Specifications
| 13455-21-5 | |
| 500 g | |
| MFCD00149929 | |
| O.O.[F-].[K+] | |
| 94.13 | |
| Reagent |
| 1 | |
| FH4KO2 | |
| MZFXHSQFQLKKFD-UHFFFAOYSA-M | |
| potassium dihydrate fluoride | |
| 99 to 102% | |
| Poly Bottle |
Chemical Identifiers
| 13455-21-5 | |
| 94.13 | |
| MZFXHSQFQLKKFD-UHFFFAOYSA-M | |
| O.O.[F-].[K+] |
| FH4KO2 | |
| MFCD00149929 | |
| potassium dihydrate fluoride |