missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride, as Dihydrate, Certified, 50% (w/v), LabChem™
$158.77 - $2182.03
Chemical Identifiers
| CAS | 7789-23-3 |
|---|---|
| Molecular Formula | FK |
| Molecular Weight (g/mol) | 58.10 |
| MDL Number | MFCD00011398 |
| InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
| Synonym | potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 |
| PubChem CID | 522689 |
| ChEBI | CHEBI:66872 |
| IUPAC Name | potassium fluoride |
| SMILES | [F-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
LC191101
|
LabChem
LC191101 |
500 mL |
Each for $158.77
|
|
|||||
|
LC191102
|
LabChem
LC191102 |
1 L |
Each for $297.80
|
|
|||||
|
LC191104
|
LabChem
LC191104 |
4 L |
Each for $591.30
|
|
|||||
|
LC191106
|
LabChem
LC191106 |
10 L |
Each for $1,333.33
|
|
|||||
|
LC191105
|
LabChem
LC191105 |
20 L |
Each for $2,182.03
|
|
|||||
Chemical Identifiers
| 7789-23-3 | |
| 58.10 | |
| NROKBHXJSPEDAR-UHFFFAOYSA-M | |
| 522689 | |
| potassium fluoride |
| FK | |
| MFCD00011398 | |
| potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
| CHEBI:66872 | |
| [F-].[K+] |
Specifications
| 7789-23-3,7732-18-5 | |
| 70.1,29.9 | |
| 500 mL | |
| KF | |
| potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
| NROKBHXJSPEDAR-UHFFFAOYSA-M | |
| potassium fluoride | |
| 522689 | |
| 58.1 | |
| Certified | |
| Poly Bottle | |
| Hydrogen fluoride | |
| Potassium Fluoride, as Dihydrate |
| Colorless | |
| Liquid | |
| FK | |
| MFCD00011398 | |
| Soluble in water | |
| [F-].[K+] | |
| 58.10 | |
| CHEBI:66872 | |
| 50% (w/v) | |
| Passes Test | |
| 1.22g/mL ±0.01 | |
| 1.22g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed or if inhaled.
GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wash exposed skin thoroughly after handling.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
Warning
Recommended Storage : Room Temperature