missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, as Dihydrate, Certified, 50% (w/v), LabChem™

Supplier:  LabChem LC191105

 View more versions of this product

Catalog No. LC191105

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Fluoride
7789-23-3,7732-18-5
70.1,29.9
20 L
KF
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
522689
58.1
Certified
Cubitainer™
Hydrogen fluoride
as Dihydrate
Colorless
Liquid
FK
MFCD00011398
Soluble in water
[F-].[K+]
58.10
CHEBI:66872
50% (w/v)
Passes Test
1.22g/mL +/- 0.01
1.22g/mL

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
potassium fluoride
FK
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
CHEBI:66872
[F-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed or if inhaled.

GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wash exposed skin thoroughly after handling.
If inhaled: Remove person to fr

Warning

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions