missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, as Dihydrate, Certified, 50% (w/v), LabChem™

Catalog No. LC191106
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC191106 10 L
LC191101 500 mL
LC191102 1 L
LC191104 4 L
LC191105 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. LC191106 Supplier LabChem Supplier No. LC191106
Only null left

Chemical Identifiers

CAS 7789-23-3 , 7732-18-5
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Color Colorless
CAS Max % 70.1, 29.9
Physical Form Liquid
Quantity 10 L
Linear Formula KF
Solubility Information Soluble in water
Formula Weight 58.1
Percent Purity 50% (w/v)
Grade Certified
Identification Passes Test
Packaging Cubitainer™
Specific Gravity 1.22g/mL ±0.01
Decomposition Information Hydrogen fluoride
Density 1.22g/mL
Chemical Name or Material Potassium Fluoride, as Dihydrate
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Harmful if swallowed or if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing mist, vapors, spray.
Do not eat, drink or smoke when using this product.
Use only outdoors or in a well-ventilated area.
Wash exposed skin thoroughly after handling.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature