missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride, as Dihydrate, Certified, 50% (w/v), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 70.1, 29.9 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Linear Formula | KF |
| Solubility Information | Soluble in water |
| Formula Weight | 58.1 |
| Percent Purity | 50% (w/v) |
| Grade | Certified |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed or if inhaled. |
| Precautionary Statement | GHS P Statement Avoid breathing mist, vapors, spray. Do not eat, drink or smoke when using this product. Use only outdoors or in a well-ventilated area. Wash exposed skin thoroughly after handling. If inhaled: Remove person to fresh air and keep comfortable for breathing. Call a poison center/doctor if you feel unwell. If swallowed: Rinse mouth. Call a poison center/doctor if you feel unwell. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Room Temperature |