missing translation for 'onlineSavingsMsg'
Learn More

Potassium Fluoride, 50% (w/w), pH 8.3, Ricca Chemical

Supplier:  Ricca Chemical Company 61051

 View more versions of this product

Catalog No. 6105-1


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Fluoride
70.0,49.0
FK
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
[F-].[K+]
58.10
CHEBI:66872
Laboratory
Natural Poly Bottle
4 L
7789-23-3,7732-18-5
73.0,51.0
MFCD00011398
NROKBHXJSPEDAR-UHFFFAOYSA-M
potassium fluoride
522689
50%
50% (w/w) Aqueous Solution
8.3

Chemical Identifiers

7789-23-3
58.10
NROKBHXJSPEDAR-UHFFFAOYSA-M
522689
potassium fluoride
FK
MFCD00011398
potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
CHEBI:66872
[F-].[K+]
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions