missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Fluoride, 40% (w/v), Ricca Chemical
For APHA/ASTM/EPA Testing
$623.09 - $1315.66
Chemical Identifiers
| Molecular Formula | FK |
|---|---|
| MDL Number | MFCD00011398 |
| InChI Key | NROKBHXJSPEDAR-UHFFFAOYSA-M |
| Synonym | potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 |
| PubChem CID | 522689 |
| ChEBI | CHEBI:66872 |
| IUPAC Name | potassium fluoride |
| SMILES | [F-].[K+] |
Chemical Identifiers
| FK | |
| NROKBHXJSPEDAR-UHFFFAOYSA-M | |
| 522689 | |
| potassium fluoride |
| MFCD00011398 | |
| potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568 | |
| CHEBI:66872 | |
| [F-].[K+] |
Specifications
| 7732-18-5 | |
| 40% | |
| Natural Poly Bottle | |
| 8.2 to 8.4 | |
| Potassium Fluoride |
| 294.18 g/mol | |
| Laboratory | |
| APHA/ASTM/EPA for Dissolved Oxygen (DO) | |
| 1 L |