missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferrocyanide, Certified, LabChem™

Catalog No. p-5388171
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC190601 500 mL
LC190602 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC190601 Supplier LabChem Supplier No. LC190601
Only null left

Chemical Identifiers

CAS 14459-95-1 , 7732-18-5
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Yellow
CAS Max % 89.2, 10.8
Physical Form Liquid
Quantity 500 mL
Linear Formula K4Fe(CN)6·3H2O
Solubility Information Soluble in water
Formula Weight 422.39
Grade Certified
Identification Passes Test
Packaging Amber Poly
Specific Gravity 1.06g/mL
Decomposition Information Hydrogen cyanide; Carbon monoxide; Carbon dioxide; Sulfur compounds
Density 1.06g/mL
Chemical Name or Material Potassium Ferrocyanide
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature