missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferricyanide, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
P1285, 13746-66-2, K3Fe(CN)6
$276.89 - $12231.36
Chemical Identifiers
| CAS | 13746-66-2 |
|---|---|
| Molecular Formula | C6FeK3N6 |
| Molecular Weight (g/mol) | 329.25 |
| MDL Number | MFCD00011392 |
| InChI Key | QMTKJUMXUDIUAQ-UHFFFAOYSA-N |
| IUPAC Name | tripotassium hexacyanoirontris(ylium) |
| SMILES | [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605539
|
Spectrum Chemical Mfg Cor
P128525GM |
25 g | Amber Glass Bottle |
Each for $276.89
|
|
||||
|
18605540
|
Spectrum Chemical Mfg Cor
P1285125GM |
125 g | Amber Glass Bottle |
Each for $537.32
|
|
||||
|
18605542
|
Spectrum Chemical Mfg Cor
P1285500GM |
500 g | Amber Glass Bottle |
Each for $885.96
|
|
||||
|
18605541
|
Spectrum Chemical Mfg Cor
P1285125GMCS6 |
6 x 125 g | Amber Glass Bottle |
Case of 6 Each for $2,481.18
|
|
||||
|
18605544
|
Spectrum Chemical Mfg Cor
P128525KG |
2.5 kg | Amber Glass Bottle |
Each for $3,164.61
|
|
||||
|
18605543
|
Spectrum Chemical Mfg Cor
P1285500GMCS6 |
6 x 500 g | Amber Glass Bottle |
Case of 6 Each for $4,779.39
|
|
||||
|
18605545
|
Spectrum Chemical Mfg Cor
P128525KGCS4 |
4 x 2.5 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18605546
|
Spectrum Chemical Mfg Cor
P128512KG |
12 kg | Poly Pail |
Each for $10,494.93
|
|
||||
Description
Spectrum™ Chemical Potassium Ferricyanide, Crystal, Reagent, ACS is a chemical compound and a bright red salt. It is soluble in water and appears as a yellow-green fluorescence. Potassium ferricyanide is created by passing chlorine through a solution of potassium ferrocyanide. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
| 13746-66-2 | |
| 329.25 | |
| QMTKJUMXUDIUAQ-UHFFFAOYSA-N | |
| [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N |
| C6FeK3N6 | |
| MFCD00011392 | |
| tripotassium hexacyanoirontris(ylium) |
Specifications
| 13746-66-2 | |
| 25 g | |
| MFCD00011392 | |
| [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N | |
| 329.25 | |
| ACS |
| 1 | |
| C6FeK3N6 | |
| QMTKJUMXUDIUAQ-UHFFFAOYSA-N | |
| tripotassium hexacyanoirontris(ylium) | |
| 99% | |
| Amber Glass Bottle |