missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferricyanide, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
P1285, 13746-66-2, K3Fe(CN)6
Supplier: Spectrum Chemical Mfg Cor P1285125GMCS6
Description
Spectrum™ Chemical Potassium Ferricyanide, Crystal, Reagent, ACS is a chemical compound and a bright red salt. It is soluble in water and appears as a yellow-green fluorescence. Potassium ferricyanide is created by passing chlorine through a solution of potassium ferrocyanide. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Specifications
| 13746-66-2 | |
| 6 x 125 g | |
| MFCD00011392 | |
| [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N | |
| 329.25 | |
| Amber Glass Bottle |
| 1 | |
| C6FeK3N6 | |
| QMTKJUMXUDIUAQ-UHFFFAOYSA-N | |
| tripotassium hexacyanoirontris(ylium) | |
| 99% |
Chemical Identifiers
| 13746-66-2 | |
| 329.25 | |
| QMTKJUMXUDIUAQ-UHFFFAOYSA-N | |
| [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N |
| C6FeK3N6 | |
| MFCD00011392 | |
| tripotassium hexacyanoirontris(ylium) |