missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical

Catalog No. 18605540
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605540 125 g Amber Glass Bottle
18605539 25 g Amber Glass Bottle
18605542 500 g Amber Glass Bottle
18605541 6 x 125 g Amber Glass Bottle
18605544 2.5 kg Amber Glass Bottle
18605543 6 x 500 g Amber Glass Bottle
18605545 4 x 2.5 kg Amber Glass Bottle
18605546 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18605540 Supplier Spectrum Chemical Mfg Cor Supplier No. P1285125GM
Only null left

P1285, 13746-66-2, K3Fe(CN)6

Spectrum™ Chemical Potassium Ferricyanide, Crystal, Reagent, ACS is a chemical compound and a bright red salt. It is soluble in water and appears as a yellow-green fluorescence. Potassium ferricyanide is created by passing chlorine through a solution of potassium ferrocyanide. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
IUPAC Name tripotassium hexacyanoirontris(ylium)
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle