missing translation for 'onlineSavingsMsg'
Learn More

Potassium dichromate concentrate, For 1L standard solution, 1/60 M K2Cr2O7 (0.1N), Honeywell Fluka™

Catalog No. 60046694
Change view
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Packaging:
Ampoule
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60046694 6 x 1 Ea. Ampoule
6001213 1 Pc. Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60046694 Supplier Honeywell Chemicals Supplier No. 381006X1EA
Only null left

for 1L standard solution, 1/60 M K2Cr2O7 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
MDL Number MFCD00011367
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Boiling Point 100°C
CAS Max % 5.7%, >94.0000%
CAS Min % >94.0000%
Physical Form Concentrate
Quantity 6 x 1 Ea.
Linear Formula K2Cr2O7
UN Number UN2922
Formula Weight 294.18
Packaging Ampoule
Density 1.030 g/cm3
Chemical Name or Material Potassium dichromate concentrate, For 1L standard solution, 1/60 M K2Cr2O7 (0.1N)
Show More Show Less
Hazard Statement P201-P260-P280-P284-P305 + P351 + P338-P308 + P313
ShelfLife 1260 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Corrosive liquids, toxic, n.o.s.
Recommended Storage Room Temp