missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, Certified, 0.200N ±0.001N (0.0333M), LabChem™

Catalog No. LC190001
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC190001 500 mL
LC190002 1 L
LC190004 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. LC190001 Supplier LabChem Supplier No. LC190001
Only null left

Chemical Identifiers

CAS 7778-50-9 , 7732-18-5
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.182
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Color Orange
CAS Max % 99, 1
Physical Form Liquid
Quantity 500 mL
Linear Formula K2Cr2O7
Solubility Information Soluble in water
Formula Weight 294.18
Grade Certified
Concentration or Composition (by Analyte or Components) 0.200N ±0.001N (0.0333M)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.01g/mL
Decomposition Information Contains hexavalent Chromium
Density 1.01g/mL
Chemical Name or Material Potassium Dichromate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs (blood, kidneys, liver) through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist.
Do not eat, drink or smoke when using this product.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature