missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dichromate, Certified, 0.1000N ±0.0005N (0.0167M), LabChem™

Supplier:  LabChem LC189806

 View more versions of this product

Catalog No. LC189806


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Dichromate
Orange
Liquid
Cr2K2O7
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
KMUONIBRACKNSN-UHFFFAOYSA-N
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
24502
294.18
0.1000N ±0.0005N (0.0167M)
Cubitainer™
Contains hexavalent Chromium
7778-50-9,7732-18-5
99.5,0.5
10 L
K2Cr2O7
Soluble in water
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
294.182
CHEBI:53444
Certified
Passes Test
1g/mL
1g/mL

Chemical Identifiers

7778-50-9
294.182
potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
CHEBI:53444
[O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]
Cr2K2O7
KMUONIBRACKNSN-UHFFFAOYSA-N
24502
dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Contaminated work clothing must not be allowed out of the workplace.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions