Learn More
Potassium cyclopropyltrifluoroborate, 97%, Thermo Scientific™
$208.22 - $554.58
Chemical Identifiers
| CAS | 1065010-87-8 |
|---|---|
| Molecular Formula | C3H5BF3K |
| Molecular Weight (g/mol) | 147.98 |
| InChI Key | CFMLURFHOSOXRC-UHFFFAOYSA-N |
| Synonym | potassium cyclopropyltrifluoroborate, potassium cyclopropyltrifluoroboranuide, potassium cyclopropyltrifluororate, potassium cyclopropytrifluoroborate, potassium cycloproyltrifluoroborate, potassiumcyclopropyltrifluoroborate, potassium cyclopropyl-trifluoroborate, cyclopropyltrifluoroborate potassium salt, potassium;cyclopropyl trifluoro boranuide, potassium ion cyclopropyltrifluoroboranuide |
| PubChem CID | 23697338 |
| IUPAC Name | potassium;cyclopropyl(trifluoro)boranuide |
| SMILES | [B-](C1CC1)(F)(F)F.[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC444870010
|
N/A | 1g | Glass bottle |
Each for $208.22
|
|
||||
|
AC444870050
|
N/A | 5g | Glass bottle |
Each for $554.58
|
|
||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 1065010-87-8 | |
| 147.98 | |
| potassium cyclopropyltrifluoroborate, potassium cyclopropyltrifluoroboranuide, potassium cyclopropyltrifluororate, potassium cyclopropytrifluoroborate, potassium cycloproyltrifluoroborate, potassiumcyclopropyltrifluoroborate, potassium cyclopropyl-trifluoroborate, cyclopropyltrifluoroborate potassium salt, potassium;cyclopropyl trifluoro boranuide, potassium ion cyclopropyltrifluoroboranuide | |
| potassium;cyclopropyl(trifluoro)boranuide |
| C3H5BF3K | |
| CFMLURFHOSOXRC-UHFFFAOYSA-N | |
| 23697338 | |
| [B-](C1CC1)(F)(F)F.[K+] |
Specifications
| 1065010-87-8 | |
| C3H5BF3K | |
| CFMLURFHOSOXRC-UHFFFAOYSA-N | |
| potassium;cyclopropyl(trifluoro)boranuide | |
| 23697338 | |
| 97% |
| 97% | |
| potassium cyclopropyltrifluoroborate, potassium cyclopropyltrifluoroboranuide, potassium cyclopropyltrifluororate, potassium cyclopropytrifluoroborate, potassium cycloproyltrifluoroborate, potassiumcyclopropyltrifluoroborate, potassium cyclopropyl-trifluoroborate, cyclopropyltrifluoroborate potassium salt, potassium;cyclopropyl trifluoro boranuide, potassium ion cyclopropyltrifluoroboranuide | |
| [B-](C1CC1)(F)(F)F.[K+] | |
| 147.98 | |
| 147.98 | |
| Potassium cyclopropyltrifluoroborate |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only