missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyclopropyltrifluoroborate, 97%

Catalog No. AC444870050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC444870050 5 g Glass bottle
AC444870010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC444870050 Supplier Thermo Scientific Chemicals Supplier No. 444870050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1065010-87-8 | C3H5BF3K | 147.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1065010-87-8
Molecular Formula C3H5BF3K
Molecular Weight (g/mol) 147.98
InChI Key CFMLURFHOSOXRC-UHFFFAOYSA-N
Synonym potassium cyclopropyltrifluoroborate, potassium cyclopropyltrifluoroboranuide, potassium cyclopropyltrifluororate, potassium cyclopropytrifluoroborate, potassium cycloproyltrifluoroborate, potassiumcyclopropyltrifluoroborate, potassium cyclopropyl-trifluoroborate, cyclopropyltrifluoroborate potassium salt, potassium;cyclopropyl trifluoro boranuide, potassium ion cyclopropyltrifluoroboranuide
PubChem CID 23697338
IUPAC Name potassium;cyclopropyl(trifluoro)boranuide
SMILES [B-](C1CC1)(F)(F)F.[K+]

Specifications

Melting Point 348°C to 350°C
Color White
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 147.98
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Potassium cyclopropyltrifluoroborate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only