accessibility menu, dialog, popup

UserName

CAS RN 1065010-87-8

IUPAC Name: potassium cyclopropyltrifluoroboranuide
Synonyms: potassium cyclopropyltrifluoroboranuide
Molecular Weight (g/mol): 147.98
Molecular Formula: C3H5BF3K
InChi Key: CFMLURFHOSOXRC-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)