missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide, 10% (w/v), Ricca Chemical

Catalog No. R60400001B
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R60400001B 1 L Amber Poly Bottle
604016 500 mL Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R60400001B Supplier Ricca Chemical Company Supplier No. R60400001B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 151-50-8 , 7732-18-5
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Color Colorless
Assay Percent Range 10% (w/v)
Percent Purity 10%
Grade EP
Odor Almond-like
Packaging Amber Poly Bottle
Solvent or Matrix Aqueous Solution
pH Alkaline
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Potassium Cyanide, 10% (w/v) Aqueous Solution
Show More Show Less