missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide, 10% (w/v), Ricca Chemical

Catalog No. 604016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
604016 500 mL Amber Poly Bottle
R60400001B 1 L Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 604016 Supplier Ricca Chemical Company Supplier No. 604016

Chemical Identifiers

CAS 151-50-8 , 7732-18-5
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Color Colorless
CAS Min % 89.0, 8.91
CAS Max % 93.0, 9.28
Percent Purity 10%
Grade Laboratory
Packaging Amber Poly Bottle
Solvent or Matrix Aqueous Solution
pH Alkaline
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 10% (w/v)
Chemical Name or Material Potassium Cyanide
Show More Show Less