missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride Reference Solution C, 1408.8 μS/cm at 25°C, 0.01 M, Ricca Chemical

Supplier:  Ricca Chemical Company 589516

 View more versions of this product

Catalog No. 5895-16


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Chloride Reference Solution C
Natural Poly Bottle
98.0,0.07
Liquid
MFCD00011360
Approximately 7
Conductivity Standard
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
500 mL
CHEBI:32588
1408.8μS/cm (1408.8μmho/cm) at 25°C
7447-40-7,7732-18-5
100.0,0.07
ClK
Colorless
100°C
Conductivity Standard
[Cl-].[K+]
74.55
4873
0.01M

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions