missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals
$214.82 - $630.06
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19142701
|
MP Biomedicals Inc
0219142701 |
1 kg |
Each for $214.82
|
|
|||||
|
ICN19142705
|
MP Biomedicals Inc
0219142705 |
5 kg |
Each for $630.06
|
|
|||||
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| potassium chloride |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 | |
| [Cl-].[K+] |
Specifications
| 7447-40-7 | |
| 1 kg | |
| MFCD00011360 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| 74.55 | |
| Potassium Chloride |
| Crystalline | |
| ClK | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| ACS Reagent |
Safety and Handling
Recommended Storage : Room Temperature