missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals

Supplier:  MP Biomedicals Inc 0219142701

 View more versions of this product

Catalog No. ICN19142701


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Chloride
Crystalline
ClK
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
[Cl-].[K+]
74.55
CHEBI:32588
ACS Reagent
7447-40-7
1 kg
MFCD00011360
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
4873
74.55

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
4873
potassium chloride
ClK
MFCD00011360
potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
CHEBI:32588
[Cl-].[K+]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions