missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Crystal, FCC, 99%, Spectrum™ Chemical
P1254, 7447-40-7, KCl
$411.37 - $3641.43
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605496
|
Spectrum Chemical Mfg Cor
P1254500GM |
500 g | Poly Bottle |
Each for $411.37
|
|
||||
|
18605497
|
Spectrum Chemical Mfg Cor
P125425KG |
2.5 kg | Poly Pail |
Each for $1,188.67
|
|
||||
|
18605498
|
Spectrum Chemical Mfg Cor
P125412KG |
12 kg | Poly Pail |
Each for $2,003.90
|
|
||||
|
18605499
|
Spectrum Chemical Mfg Cor
P125445KGBL |
45 kg | Fiber Drum |
Each for $3,641.43
|
|
||||