missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Crystal, FCC, 99%, Spectrum™ Chemical

P1254, 7447-40-7, KCl

Supplier:  Spectrum Chemical Mfg Cor P125445KGBL

 View more versions of this product

Catalog No. 18605499


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Chloride, Crystal, FCC is used as a flavor enhancer, flavoring agent, nutrient supplement, pH control agent, and stabilizer or thickener. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

7447-40-7
45 kg
MFCD00011360
[Cl-].[K+]
74.55
FCC
Fiber Drum
1
ClK
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
99%
0.01

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
[Cl-].[K+]
ClK
MFCD00011360
potassium chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions