missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$7232.56 - $7232.56
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| Synonym | potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| SMILES | [Cl-].[K+] |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 |
Specifications
| 7447-40-7 | |
| 2.5 kg | |
| MFCD00011360 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium;chloride | |
| 4873 | |
| 74.55 | |
| Poly Bottle | |
| Potassium Chloride |
| Crystals | |
| ClK | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| BAKER ANALYZED™ ACS Reagent | |
| 1L = 1.99kg |