missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02003738
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02003738 2.5 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02003738 Supplier Avantor Supplier No. 304005
Only null left

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Physical Form Crystals
Quantity 2.5 kg
IUPAC Name potassium;chloride
Formula Weight 74.55
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.99kg
Chemical Name or Material Potassium Chloride