missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride Conductivity Standard, 20,000 μS/cm (20 mS/cm) at 25°C, Ricca Chemical
$315.23 - $315.23
Chemical Identifiers
| CAS | 7447-40-7 |
|---|---|
| Molecular Formula | ClK |
| Molecular Weight (g/mol) | 74.55 |
| MDL Number | MFCD00011360 |
| InChI Key | WCUXLLCKKVVCTQ-UHFFFAOYSA-M |
| PubChem CID | 4873 |
| ChEBI | CHEBI:32588 |
| IUPAC Name | potassium chloride |
| SMILES | [Cl-].[K+] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
R58882004A
|
Ricca Chemical Company
R58882004A |
4 L | Natural Poly Bottle |
Each for $315.23
|
|
||||
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Specifications
| Natural Poly Bottle | |
| Odorless | |
| Liquid | |
| MFCD00011360 | |
| Approximately 7 | |
| Conductivity Standard | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| Water | |
| 74.55 | |
| 4873 | |
| Potassium Chloride Conductivity Standard, 20 mS/cm (20, 000μmho/cm) at 25°C |
| 7447-40-7,7732-18-5 | |
| 20.0 ± 0.1 mS/cm at 25°C | |
| ClK | |
| Colorless | |
| 100°C | |
| Conductivity Standard | |
| [Cl-].[K+] | |
| potassium chloride | |
| 4 L | |
| CHEBI:32588 |