missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, 99+%, ACS Reagent
CAS: 7447-40-7 | ClK | 74.55 g/mol
Supplier: Thermo Scientific Chemicals 424090050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 1420.0°C | |
| 770.0°C | |
| White | |
| Crystalline Powder and/or Crystals | |
| 99.0 to 100.5% (Argentometry) | |
| KCl | |
| 06,480 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| 74.54 | |
| ACS Reagent |
| Potassium chloride | |
| 7447-40-7 | |
| 5.4 to 8.6 (5% soln. at 25°C) | |
| 5 kg | |
| ClK | |
| MFCD00011360 | |
| 15, 7742 | |
| Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| ≥99% | |
| Plastic Bottle |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 231-211-8
RTECSNumber : TS8050000
TSCA : TSCA
RUO – Research Use Only