missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, 99+%, ACS Reagent

Catalog No. p-43004
Change view
Click to view available options
Quantity:
25 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424090250 25 g Glass bottle
AC424090010 1 kg Plastic bottle
AC424090050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC424090250 Supplier Thermo Scientific Chemicals Supplier No. 424090250
Only null left

CAS: 7447-40-7 | ClK | 74.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1420.0°C
Melting Point 770.0°C
Color White
pH 5.4 to 8.6 (5% soln. at 25°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 25 g
Assay Percent Range 99.0 to 100.5% (Argentometry)
Linear Formula KCl
Fieser 06,480
Merck Index 15, 7742
Solubility Information Solubility in water: 340g/L (20°C). Other solubilities: slightly soluble in alcohol, insoluble in ether and acetone
IUPAC Name potassium;chloride
Formula Weight 74.54
Percent Purity ≥99%
Grade ACS Reagent
Packaging Glass bottle
Chemical Name or Material Potassium chloride
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 231-211-8
RTECSNumber TS8050000
TSCA TSCA

RUO – Research Use Only