missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, 4 M, saturated with Silver Chloride, Electrode Filling Solution, Ricca Chemical
Supplier: Ricca Chemical Company 592016
Specifications
| Potassium Chloride | |
| Electrode Filling Solution | |
| ClK | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| [Cl-].[K+] | |
| 74.55 | |
| CHEBI:32588 | |
| Natural Poly Bottle | |
| 500 mL | |
| 4Molar,saturated with Silver Chloride |
| Colorless | |
| 7447-40-7,7761-88-8,7732-18-5 | |
| MFCD00011360 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 4873 | |
| Laboratory | |
| ∼7 | |
| Liquid |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| 4873 | |
| potassium chloride |
| ClK | |
| MFCD00011360 | |
| potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs | |
| CHEBI:32588 | |
| [Cl-].[K+] |