missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Chloride, 3.0M (3.0N), LabChem™
Specifications
Specifications
| Color | Colorless |
| CAS Max % | 80, 20 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Linear Formula | KCl |
| Solubility Information | Soluble in water |
| Formula Weight | 74.55 |
| Grade | Certified |
| Concentration or Composition (by Analyte or Components) | 3.00M ± 0.05M |
| Identification | Passes Test |
| Show More |
Safety and Handling
| Recommended Storage | Room Temperature |