missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 0.0145M, Conductivity Standard

Catalog No. p-7052159
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42693AP 500 mL
AA42693K2 1 L
AA42693K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA42693AP Supplier Thermo Scientific Chemicals Supplier No. 042693AP
Only null left

CAS: 7447-40-7 | ClK | 74.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Odor Odorless
Physical Form Liquid
Concentration 0.0145 M
IUPAC Name potassium chloride
Quantity 500 mL
Formula Weight 74.55
Chemical Name or Material Potassium chloride Conductivity Standard, 2000μmho at 25°C
EINECSNumber 231-211-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only