missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, 0.0100 M (M/100), 1412 μS/cm at 25°C, Ricca Chemical

Supplier:  Ricca Chemical Company 589032

 View more versions of this product

Catalog No. 5890-32


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Chloride
Natural Poly Bottle
0.0100Molar
ClK
Colorless
100°C
Conductivity Standard
[Cl-].[K+]
74.55
4873
1412μS/cm (1412μmho/cm) at 25°C
7447-40-7,7732-18-5
Liquid
MFCD00011360
Approximately 7
Conductivity Standard
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
potassium chloride
1 L
CHEBI:32588

Chemical Identifiers

7447-40-7
74.55
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
CHEBI:32588
[Cl-].[K+]
ClK
MFCD00011360
4873
potassium chloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions