Learn More
Potassium chloride, 0.005M, Conductivity Standard
CAS: 7447-40-7 | ClK | 74.55 g/mol
Supplier: Thermo Scientific Chemicals 042680K2
Description
Potassium chloride 0.0005M solution is used as conductive standard and as electrolyte in analytical chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium chloride Conductivity Standard | |
| 7447-40-7 | |
| Liquid | |
| 0.005 M | |
| Miscible with water | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 | |
| 74.55 |
| 718μmho at 25°C | |
| Odorless | |
| ClK | |
| MFCD00011360 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 1 L | |
| CHEBI:32588 |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Safety and Handling
EINECSNumber : 231-211-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only