missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium chloride, 0.00017M, Conductivity Standard
CAS: 7447-40-7 | ClK | 74.55 g/mol
Supplier: Thermo Scientific Chemicals 043444K2
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium chloride Conductivity Standard | |
| 7447-40-7 | |
| Liquid | |
| 0.00017 M | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| potassium chloride | |
| 1 L | |
| CHEBI:32588 |
| 25μmho at 25°C | |
| Odorless | |
| ClK | |
| MFCD00011360 | |
| [Cl-].[K+] | |
| 74.55 | |
| 4873 | |
| 74.55 |
Chemical Identifiers
| 7447-40-7 | |
| 74.55 | |
| WCUXLLCKKVVCTQ-UHFFFAOYSA-M | |
| CHEBI:32588 | |
| [Cl-].[K+] |
| ClK | |
| MFCD00011360 | |
| 4873 | |
| potassium chloride |
Safety and Handling
EINECSNumber : 231-211-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only