missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Carbonate, Anhydrous, Granular, USP, 99.5-100.5%, Spectrum™ Chemical
P1238, 584-08-7, K2CO3
$1017.63 - $11235.73
Chemical Identifiers
| CAS | 584-08-7 |
|---|---|
| Molecular Formula | CK2O3 |
| Molecular Weight (g/mol) | 138.21 |
| MDL Number | MFCD00011382 |
| InChI Key | BWHMMNNQKKPAPP-UHFFFAOYSA-L |
| IUPAC Name | dipotassium carbonate |
| SMILES | [K+].[K+].[O-]C([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18605489
|
Spectrum Chemical Mfg Cor
P123825KG |
2.5 kg | Poly Pail |
Each for $1,017.63
|
|
||||
|
18605490
|
Spectrum Chemical Mfg Cor
P123812KG |
12 kg | Poly Pail |
Each for $3,004.10
|
|
||||
|
18605491
|
Spectrum Chemical Mfg Cor
P123845KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||