missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide Solution, For Mercury, LabChem™

Supplier:  LabChem LC187851

Catalog No. LC187851


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Bromide Solution
For Mercury
72.28,27.72
KBr
1.23g/mL
Soluble in water
[K+].[Br-]
119.00
CHEBI:32030
Passes Test
1.23g/mL
Hydrogen bromide
Liquid
Colorless
7758-02-3 , 7732-18-5
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
253877
119
Poly Bottle
0.76cSt
500 mL

Chemical Identifiers

7758-02-3
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
253877
potassium bromide
BrK
MFCD00011358
potassium bromide, bromide salt of potassium, kalii bromidum, bromure de potassium, caswell no. 684, tripotassium tribromide, potassium bromide k3br3, unii-osd78555zm, ccris 6095
CHEBI:32030
[K+].[Br-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions