missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bromide, Crystal, ACS, 99%, Spectrum™ Chemical
$227.62 - $6471.29
Chemical Identifiers
| CAS | 2-3-7758 |
|---|---|
| Molecular Formula | BrK |
| Molecular Weight (g/mol) | 119.00 |
| MDL Number | MFCD00011358 |
| InChI Key | IOLCXVTUBQKXJR-UHFFFAOYSA-M |
| IUPAC Name | potassium bromide |
| SMILES | [K+].[Br-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18612504
|
Spectrum Chemical Mfg Cor
P122025GM |
25 g | Amber Glass Bottle |
Each for $227.62
|
|
||||
|
18612505
|
Spectrum Chemical Mfg Cor
P1220125GM |
125 g | Amber Glass Bottle |
Each for $410.47
|
|
||||
|
18612506
|
Spectrum Chemical Mfg Cor
P1220500GM |
500 g | Amber Glass Bottle |
Each for $729.62
|
|
||||
|
18612507
|
Spectrum Chemical Mfg Cor
P122025KG |
2.5 kg | Amber Glass Bottle |
Each for $1,308.59
|
|
||||
|
18612508
|
Spectrum Chemical Mfg Cor
P122012KG |
12 kg | Poly Pail |
Each for $2,734.32
|
|
||||
|
18612509
|
Spectrum Chemical Mfg Cor
P122025KGBL |
25 kg | Poly Pail |
Each for $4,931.65
|
|
||||
|
18612510
|
Spectrum Chemical Mfg Cor
P122050KGBL |
50 kg | Fiber Drum |
Each for $6,471.29
|
|
||||
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.