missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P1220500GM

 View more versions of this product

Catalog No. 18612506


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-3-7758
5.0 to 8.8
BrK
IOLCXVTUBQKXJR-UHFFFAOYSA-M
potassium bromide
99%
Amber Glass Bottle
1
500 g
MFCD00011358
[K+].[Br-]
119.00
ACS

Chemical Identifiers

2-3-7758
119.00
IOLCXVTUBQKXJR-UHFFFAOYSA-M
[K+].[Br-]
BrK
MFCD00011358
potassium bromide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.