missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bromide, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18612504
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612504 25 g Amber Glass Bottle
18612505 125 g Amber Glass Bottle
18612506 500 g Amber Glass Bottle
18612507 2.5 kg Amber Glass Bottle
18612508 12 kg Poly Pail
18612509 25 kg Poly Pail
18612510 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18612504 Supplier Spectrum Chemical Mfg Cor Supplier No. P122025GM

Chemical Identifiers

CAS 2-3-7758
Molecular Formula BrK
Molecular Weight (g/mol) 119.00
MDL Number MFCD00011358
InChI Key IOLCXVTUBQKXJR-UHFFFAOYSA-M
IUPAC Name potassium bromide
SMILES [K+].[Br-]

Specifications

CAS Max % 1
pH 5.0 to 8.8
Quantity 25 g
Percent Purity 99%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.