missing translation for 'onlineSavingsMsg'
Learn More

Potassium Borohydride 98.0+%, TCI America™

Supplier:  TCI America P1681100G

 View more versions of this product

Catalog No. P1681100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Potassium Borohydride
White
Crystalline Powder
BH4K
1870
ICRGAIPBTSPUEX-UHFFFAOYSA-N
potassium boranuide
22892188
≥98.0% (V)
500°C
13762-51-1
100 g
MFCD00011396
potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
[BH4-].[K+]
53.94
53.94

Chemical Identifiers

13762-51-1
53.94
ICRGAIPBTSPUEX-UHFFFAOYSA-N
22892188
[BH4-].[K+]
BH4K
MFCD00011396
potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
potassium boranuide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-0587

RTECSNumber : TS7525000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions