missing translation for 'onlineSavingsMsg'
Learn More

Potassium Borohydride 98.0+%, TCI America™

$38.51 - $107.80

Chemical Identifiers

CAS 13762-51-1
Molecular Formula BH4K
Molecular Weight (g/mol) 53.94
MDL Number MFCD00011396
InChI Key ICRGAIPBTSPUEX-UHFFFAOYSA-N
Synonym potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
PubChem CID 22892188
IUPAC Name potassium boranuide
SMILES [BH4-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P168125G
View Documents
TCI America
P168125G
25 g
Each for $38.51
Only null left
 
P1681100G
View Documents
TCI America
P1681100G
100 g
Each for $107.80
Only null left
 
Description

Description

Specifications

Chemical Identifiers

13762-51-1
53.94
ICRGAIPBTSPUEX-UHFFFAOYSA-N
22892188
[BH4-].[K+]
BH4K
MFCD00011396
potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
potassium boranuide

Specifications

500°C
13762-51-1
25 g
MFCD00011396
potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
[BH4-].[K+]
53.94
53.94
Potassium Borohydride
White
Crystalline Powder
BH4K
1870
ICRGAIPBTSPUEX-UHFFFAOYSA-N
potassium boranuide
22892188
≥98.0% (V)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (1)-0587

RTECSNumber : TS7525000

TSCA : Yes

SDS
Documents

Documents