missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bitartrate, Powder, 99.0 -101.0%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P120512KG

 View more versions of this product

Catalog No. 18612487


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

868-14-4
12 kg
Reagent
1
99 to 101%
Poly Pail

Chemical Identifiers

868-14-4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.