missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bitartrate, Powder, 99.0 -101.0%, Spectrum™ Chemical

Catalog No. p-7166038
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612484 125 g Poly Bottle
18612485 500 g Poly Bottle
18612486 2.5 kg Poly Bottle
18612487 12 kg Poly Pail
18612488 25 kg Fiber Drum
18612489 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18612484 Supplier Spectrum Chemical Mfg Cor Supplier No. P1205125GM
Only null left

Chemical Identifiers

CAS 868-14-4

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 99 to 101%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.