missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
CAS: 40949-94-8 | C6H18KNSi2 | 199.485 g/mol
$159.71 - $940.40
Specifications
| Chemical Name or Material | Potassium bis(trimethylsilyl)amide |
|---|---|
| Color | Yellow |
| Name Note | 0.5M solution in toluene |
| CAS | 40949-94-8,108-88-3 |
| Molecular Formula | C6H18KNSi2 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC418231000
|
Thermo Scientific Chemicals
418231000 |
100 mL | AcroSeal™ Glass bottle |
Each for $159.71
|
|
||||
|
AC418238000
|
Thermo Scientific Chemicals
418238000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $940.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium bis(trimethylsilyl)amide | |
| 0.5M solution in toluene | |
| C6H18KNSi2 | |
| MFCD00010330 | |
| 0.8760g/mL | |
| IUBQJLUDMLPAGT-UHFFFAOYSA-N | |
| potassium;bis(trimethylsilyl)azanide | |
| 3251421 | |
| 0.6 to 0.8 M (Total base) |
| Yellow | |
| 40949-94-8,108-88-3 | |
| [(CH3)3Si]2NK | |
| 0.6 to 0.8M (Total base) | |
| potassium bis trimethylsilyl amide, khmds, potassium hexamethyldisilazide, hexamethyldisilazane potassium salt, potassium bis trimethylsilyl azanide, potassium bis trimethylsilyl amide 1m sol. in thf, potassiumbis trimethylsilyl amide, hexamethyldislazane potassium salt, 1,1,1,3,3,3-hexamethyldisilazane potassium salt, potassium bis-trimethylsilylamide | |
| C[Si](C)(C)[N-][Si](C)(C)C.[K+] | |
| 199.485 | |
| 199.49 | |
| Liquid |
RUO – Research Use Only