missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
CAS: 40949-94-8 | C6H18KNSi2 | 199.485 g/mol
Supplier: Thermo Scientific Chemicals 418231000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Potassium bis(trimethylsilyl)amide | |
| 0.5M solution in toluene | |
| C6H18KNSi2 | |
| MFCD00010330 | |
| 0.8760g/mL | |
| Solubility in water: reacts. | |
| C[Si](C)(C)[N-][Si](C)(C)C.[K+] | |
| 199.485 | |
| 199.49 | |
| AcroSeal™ Glass bottle | |
| 100 mL | |
| Liquid |
| Yellow | |
| 40949-94-8,108-88-3 | |
| [(CH3)3Si]2NK | |
| 0.6 to 0.8M (Total base) | |
| potassium bis trimethylsilyl amide, khmds, potassium hexamethyldisilazide, hexamethyldisilazane potassium salt, potassium bis trimethylsilyl azanide, potassium bis trimethylsilyl amide 1m sol. in thf, potassiumbis trimethylsilyl amide, hexamethyldislazane potassium salt, 1,1,1,3,3,3-hexamethyldisilazane potassium salt, potassium bis-trimethylsilylamide | |
| IUBQJLUDMLPAGT-UHFFFAOYSA-N | |
| potassium;bis(trimethylsilyl)azanide | |
| 3251421 | |
| 0.6 to 0.8 M (Total base) | |
| 0.876 | |
| −21°C |
RUO – Research Use Only