missing translation for 'onlineSavingsMsg'
Learn More

Potassium Biiodate, 0.0375 N, Ricca Chemical

Supplier:  Ricca Chemical Company R58760001C

Quantity 1 L
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. R58760001C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

13455-24-8 , 7732-18-5
389.909
potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
potassium;iodic acid;iodate
HI2KO6
ACAYDTMSDROWHW-UHFFFAOYSA-M
23700942
OI(=O)=O.[O-]I(=O)=O.[K+]

Specifications

Potassium Biiodate
98.0, 0.12
HI2KO6
ACAYDTMSDROWHW-UHFFFAOYSA-M
potassium;iodic acid;iodate
23700942
1 L
13455-24-8 , 7732-18-5
100.0, 0.12
potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
OI(=O)=O.[O-]I(=O)=O.[K+]
389.909
Amber Glass Bottle
0.0375Normal
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions