missing translation for 'onlineSavingsMsg'
Learn More

Potassium Biiodate, 0.0375 N, Ricca Chemical

Chemical Identifiers

CAS 13455-24-8 , 7732-18-5
Molecular Formula HI2KO6
Molecular Weight (g/mol) 389.909
InChI Key ACAYDTMSDROWHW-UHFFFAOYSA-M
Synonym potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
PubChem CID 23700942
IUPAC Name potassium;iodic acid;iodate
SMILES OI(=O)=O.[O-]I(=O)=O.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
R5876000500
View Documents Promotion Details
N/A 500 mL Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
R58760001C
View Documents Promotion Details
N/A 1 L Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

13455-24-8 , 7732-18-5
389.909
potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
potassium;iodic acid;iodate
HI2KO6
ACAYDTMSDROWHW-UHFFFAOYSA-M
23700942
OI(=O)=O.[O-]I(=O)=O.[K+]

Specifications

13455-24-8 , 7732-18-5
100.0, 0.12
potassium hydrogen diiodate, potassium biiodate, kaliumhydrogendijodat, iodic acid hio3 , potassium salt 2:1, potassium biiodate solution, potassium iodic acid iodate, io3.k.hio3, potassium ion iodic acid iodate, potassium hydrogen iodate powder, potassium hydrogen diiodate, p.a
OI(=O)=O.[O-]I(=O)=O.[K+]
389.909
0.0375Normal
98.0, 0.12
HI2KO6
ACAYDTMSDROWHW-UHFFFAOYSA-M
potassium;iodic acid;iodate
23700942
Potassium Biiodate
Videos
SDS
Documents

Documents