missing translation for 'onlineSavingsMsg'
Learn More

Potassium Acetate, Granular, U.S.P., J.T. Baker™

$6090.89 - $27396.11

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
02003704
View Documents
Avantor
291401
500 g Poly Bottle
Case of 4 Each for $6,090.89
Only null left
 
02003703
View Documents
Avantor
291405
2.5 kg Poly Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

127-08-2
98.142
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]
C2H3KO2
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate

Specifications

127-08-2
500 g
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CC(=O)[O-].[K+]
98.142
CHEBI:32029
USP
1L = 1.57kg
Granules
C2H3KO2
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.14
Poly Bottle
Potassium Acetate
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'